missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HS2ST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567369
This item is not returnable.
View return policy
Description
Immunogen sequence: GATELYRTGKK SHLRKTTEKK LPTKQTIAKL QQSDIWKMEN EFYEFALEQF QFIRAHAVRE KDGDLYILAQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.
Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. This gene encodes a member of the heparan sulfate biosynthetic enzyme family that transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of this gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation. Two alternatively spliced transcript variants that encode different proteins have been found for this gene.
Specifications
| HS2ST1 | |
| Polyclonal | |
| Unconjugated | |
| HS2ST1 | |
| 2OST; 2-OST; 2-O-sulfotransferase; AW214369; dJ604K5.2; heparan sulfate 2-O-sulfotransferase 1; Hs2st; Hs2st1; KIAA0448; mKIAA0448 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7LGA3 | |
| HS2ST1 | |
| Recombinant Human HS2ST1. Recombinant protein control fragment (Product #RP-108026). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction