missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HS1BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | HS1BP3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18338636
|
Novus Biologicals
NBP3-04506-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311836
|
Novus Biologicals
NBP3-04506-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HS1BP3 Polyclonal specifically detects HS1BP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HS1BP3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ14249ETM2, HCLS1 binding protein 3, HCLS1-binding protein 3, HS1-binding protein 3, HS1-BP3, HSP1BP-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HS1BP3 (NP_071905.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 64342 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title