missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRSP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531352
This item is not returnable.
View return policy
Description
Recommended positive controls: HCT116. Predicted reactivity: Mouse (81%), Rat (86%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Endoribonuclease responsible for the inhibition of the translation by cleaving mRNA. Inhibits cell-free protein synthesis. Cleaves phosphodiester bonds only in single-stranded RNA.
Specifications
| HRSP12 | |
| Polyclonal | |
| Unconjugated | |
| RIDA | |
| 14.5 kDa translational inhibitor protein; 2-iminobutanoate/2-iminopropanoate deaminase; Heat-responsive protein 12; hp14.5; HR12; HRP12; HRSP12; Liver perchloric acid-soluble protein; L-PSP; P14.5; perchloric acid soluble protein; perchloric acid-soluble protein; perchrolic acid soluble protein; Psp; PSP1; Reactive intermediate imine deaminase A homolog; reactive intermediate/imine deaminase A homolog; ribonuclease UK114; RIDA; rp14.5; Translation inhibitor L-PSP ribonuclease; translational inhibitor p14.5; translational inhibitor protein p14.5; UK114; UK114 antigen homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10247 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P52758 | |
| RIDA | |
| Full length human HRSP12 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction