missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HRP Conjugate Stabilizer/Diluent

Applications: Western Blot

Brand:  Novus Biologicals™ 901-0005

Additional Details : Weight : 0.00970kg

Product Code. 18064004

  • £351.92 / 50mL
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4°C. Do not freeze.
50 mL
HRP Conjugate Stabilizer/Diluent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers