missing translation for 'onlineSavingsMsg'
Learn More

HRP Antibody (HP-03), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18429214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429214 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429214 Supplier Novus Biologicals Supplier No. NB500420MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HRP Monoclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRP
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone HP-03
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3
Host Species Mouse
Immunogen Horseradish peroxidase (Grade I).
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.