missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRP Antibody (2H11cc), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
£428.00
Specifications
| Antigen | HRP |
|---|---|
| Clone | 2H11cc |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 750 |
Description
HRP Monoclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| HRP | |
| ELISA | |
| Alexa Fluor 750 | |
| Mouse | |
| Non-species specific | |
| EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3 | |
| IgG2b | |
| Protein A purified |
| 2H11cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified peroxidase from horseradish. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title