missing translation for 'onlineSavingsMsg'
Learn More

HRP-2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18415475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415475 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415475 Supplier Novus Biologicals Supplier No. NBP281994JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HRP-2 Polyclonal antibody specifically detects HRP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRP-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias HDGF Like 2, HDGF2, HDGF-2, HDGFL2, HDGF-Related Protein 2, HDGFRP2, Hepatoma-derived growth factor 2, Hepatoma-Derived Growth Factor Related Protein 2, Hepatoma-Derived Growth Factor-Related Protein 2, HRP2
Host Species Rabbit
Immunogen HRP-2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human HRP-2. The immunogen is located within amino acids 50 - 100 of HRP-2. Amino Acid Squence: DKDKYGKPNKRKGFNEGL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84717
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.