Learn More
Hrk Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76414MFV500
Product Code. 18455016
  • £470.00 / 0.10mL
Description
Hrk Polyclonal antibody specifically detects Hrk in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hrk
Polyclonal
Western Blot, ELISA
activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5
Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk. Amino Acid Squence: MWRRRARSRRAPAPG
0.1 mL
Neuroscience, Signal Transduction
8739
Store at 4C in the dark.
IgG
Western Blot, ELISA
mFluor Violet 500 SE
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse
Purified