missing translation for 'onlineSavingsMsg'
Learn More

Hrk Antibody, DyLight 650, Novus Biologicals™

Product Code. 18427816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427816 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427816 Supplier Novus Biologicals Supplier No. NBP176414C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hrk Polyclonal antibody specifically detects Hrk in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hrk
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk. Amino Acid Squence: MWRRRARSRRAPAPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8739
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.