missing translation for 'onlineSavingsMsg'
Få mere at vide

Hrk Antibody, DyLight 550, Novus Biologicals™

Artikelnummer. 18402348 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18402348 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18402348 Leverandør Novus Biologicals Leverandørnr. NBP176414R

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Hrk Polyclonal antibody specifically detects Hrk in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Hrk
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk. Amino Acid Squence: MWRRRARSRRAPAPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8739
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.