missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hrk Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76414CL1
This item is not returnable.
View return policy
Description
Hrk Polyclonal antibody specifically detects Hrk in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Hrk | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Hrk. The immunogen is located within the last 50 amino acids of Hrk. Amino Acid Squence: MWRRRARSRRAPAPG | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 8739 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction