missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115166
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EIF2AK1.
EIF2AK1 (HRI) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK1, signaling is initiated by low levels of heme. The HRI gene is localized to 7p22 where its 3' end slightly overlaps the 3' end of the gene JTV1. The two genes are transcribed from opposite strands and kinase activity is induced by low levels of heme and inhibited by the presence of heme.
Specifications
| HRI | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK1 | |
| EIF2AK1; eukaryotic translation initiation factor 2 alpha kinase 1; eukaryotic translation initiation factor 2-alpha kinase 1; HCR; he; heme regulated initiation factor 2 alpha kinase; heme sensitive initiation factor 2a kinase; heme-controlled repressor; Heme-regulated eukaryotic initiation factor eIF-2-alpha kinase; heme-regulated inhibitor; heme-regulated initiation factor 2-alpha kinase; heme-regulated repressor; hemin-sensitive initiation factor 2a kinase; hemin-sensitive initiation factor 2-alpha kinase; Hri; KIAA1369; PRO1362 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15467, 27102 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9BQI3, Q9Z2R9 | |
| EIF2AK1 | |
| A synthesized peptide derived from human EIF2AK1(Accession Q9BQI3), corresponding to amino acid residues I575-K625. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction