missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRH1 Recombinant Rabbit Monoclonal Antibody (HL2783)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556745
This item is not returnable.
View return policy
Description
HRH1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene was thought to be intronless until recently. The protein encoded by this gene is an integral membrane protein and belongs to the G protein-coupled receptor superfamily. It mediates the contraction of smooth muscles, the increase in capillary permeability due to contraction of terminal venules, the release of catecholamine from adrenal medulla, and neurotransmission in the central nervous system. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| HRH1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35367 | |
| HRH1 | |
| Recombinant protein encompassing a sequence within the Intracellular domain human of HRH1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2783 | |
| Unconjugated | |
| HRH1 | |
| Bordetella pertussis induced histamine sensitization; Bphs; H1 receptor; H1R; H1-R; HH1R; Hir; hisH1; Hisr; histamine H1 receptor; Histamine H1-R; histamine receptor; histamine receptor H 1; histamine receptor H1; histamine receptor, subclass H1; Hrh1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3269 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction