missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13413-1-AP-150UL
This item is not returnable.
View return policy
Description
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene was thought to be intronless until recently. The protein encoded by this gene is an integral membrane protein and belongs to the G protein-coupled receptor superfamily. It mediates the contraction of smooth muscles, the increase in capillary permeability due to contraction of terminal venules, the release of catecholamine from adrenal medulla, and neurotransmission in the central nervous system. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| HRH1 | |
| Polyclonal | |
| Unconjugated | |
| HRH1 | |
| H1 R, H1R, HH1R, hisH1, Histamine H1 receptor, histamine receptor H1, HRH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15465, 24448, 3269 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31390, P35367, P70174 | |
| HRH1 | |
| HRH1 Fusion Protein Ag4067 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction