Learn More
Invitrogen™ HRG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597051
Description
Immunogen sequence: SPTDCSAVEP EAEKALDLIN KRRRDGYLFQ LLRIADAHLD RVENTTVYYL VLDVQESDCS VLSRKYWNDC EPPDSRRPSE IVIGQCKVIA TRHSHESQDL RVIDFNCTTS SVSSALANTK DSPVLIDFFE DTERYRKQAN KALEKYKEEN DDFASFRVDR IERVARVRGG EGTGYFVDFS VRNCPRHHFP RHPNVFGFCR ADLFYDVEAL DLESPKNLVI NCEVFDPQEH ENINGVPPHL G; Positive Samples: Mouse pancreas.
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
Specifications
| HRG | |
| Polyclonal | |
| Unconjugated | |
| HRG | |
| AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171016, 94175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99PS8, Q9ESB3 | |
| HRG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-260 of human HRG (NP_0004031). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.