Learn More
PeproTech HRG beta-1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | HRG beta-1 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17855523
|
PeproTech
500-P288BT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17885013
|
PeproTech
500-P288BT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17865013
|
Invitrogen
500-P288BT-1MG |
2 x 500 μg | |||||||
|
30054619
|
PeproTech
500-P288BT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Heregulinbeta-1. Anti-Human Heregulinbeta-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Heregulin-beta1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Heregulin-beta1 (500-P288) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Heregulin-beta1. Western Blot: To detect Human Heregulin-beta1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human Heregulin-beta1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P288BT-1MG will be provided as 2 x 500 μg
Direct ligand for ERBB3 and ERBB4 tyrosine kinase receptors. Concomitantly recruits ERBB1 and ERBB2 coreceptors, resulting in ligand-stimulated tyrosine phosphorylation and activation of the ERBB receptors. The multiple isoforms perform diverse functions such as inducing growth and differentiation of epithelial, glial, neuronal, and skeletal muscle cells; inducing expression of acetylcholine receptor in synaptic vesicles during the formation of the neuromuscular junction; stimulating lobuloalveolar budding and milk production in the mammary gland and inducing differentiation of mammary tumor cells; stimulating Schwann cell proliferation; implication in the development of the myocardium such as trabeculation of the developing heart. Isoform 10 may play a role in motor and sensory neuron development.Specifications
| HRG beta-1 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| Q02297 | |
| 3084 | |
| E.coli-derived Recombinant Human Heregulinbeta-1. | |
| Antigen affinity chromatography | |
| Nrg1 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 6030402G23Rik; acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; D230005F13Rik; GGF; GGF2; GGFII; Glial growth factor; glial growth factor 2; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); Hgl; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; Neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; Neuregulin-1; Nrg1; NRG1-IT2; Pro-neuregulin-1, membrane-bound isoform; pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; SMDF; type I neuregulin 1; type III neuregulin 1 | |
| Nrg1 | |
| Primary | |
| -20°C |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.