Learn More
HRD1/SYVN1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-13473-100UL
Beskrivelse
The HRD1/SYVN1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HRD1/SYVN1. This antibody recognizes human, mouse, rat antigen. The HRD1/SYVN1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Tekniske data
| HRD1/SYVN1 | |
| 1000 μg/mL | |
| HRD1, KIAA1810, Synovial apoptosis inhibitor 1, SYVN1 | |
| 100 μL | |
| 84447 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.