missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRD1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142539
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoforms a and b (NP_115807.1; NP_757385.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
HRD1 is a ubiquitin ligase whose expression is induced by the unfolded protein response (UPR) following endoplasmic reticulum stress. Expression of HRD1 protects cells from apoptosis by inducing degradation of abnormally processed proteins that accumulate in the endoplasmic reticulum. HRD1 is expressed in many tissues, strongly expressed in brain, pancreas, liver, kidney and skeletal muscle. It has been reported that Synoviolin/Hrd1 (expressed in rheumatoid synovium) is a novel causative factor for arthropathy by triggering synovial cell outgrowth through its anti-apoptotic effects. HRD1 contains one ring-type zinc finger.
Specifications
| HRD1 | |
| Polyclonal | |
| Unconjugated | |
| SYVN1 | |
| 1200010C09Rik; AW211966; C85322; D530017H19Rik; DER3; E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; Hrd1; HRD1 protein; KIAA1810; MGC40372; RGD1310488; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin; synoviolin 1; Syvn1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 84447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q86TM6 | |
| SYVN1 | |
| Peptide with sequence C-RALEGHERQHLE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction