missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18142-1-AP-20UL
This item is not returnable.
View return policy
Description
May play a role in the regulation of calcium sequestration or release in the SR of skeletal and cardiac muscle.Specifications
| HRC | |
| Polyclonal | |
| Unconjugated | |
| HRC | |
| HCP, HRC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15464, 3270 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23327 | |
| Hrc | |
| HRC Fusion Protein Ag12794 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction