missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRASLS2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-20931
This item is not returnable.
View return policy
Description
HRASLS2 Polyclonal specifically detects HRASLS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| HRASLS2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ20556, HRAS-like suppressor 2, PLA1/2-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54979 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_060348.1 | |
| PLAAT2 | |
| Peptide with sequence QELPYSLTSDNC corresponding to internal region according to NP_060348.1. | |
| 0.1 mg | |
| Primary | |
| This design was based on provisional sequence data. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction