missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: DGYVINIAPV DGIPASFTSA KSVFSSKALV KMQLLKDVVG NDTYRINNKY DETYPPLPVE EIIKRSEFVI GQEVAYNLLV NN Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Specifications
Specifications
| Antigen | HRASLS |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HRASLS |
| Gene Accession No. | Q9HDD0 |
| Gene Alias | A-C1; HRAS like suppressor; HRAS-like suppressor 1; HRASLS; HRASLS1; H-REV107; H-REV107 protein-related protein; HRSL1; HSD28; phospholipase A; Phospholipase A and acyltransferase 1; Phospholipid-metabolizing enzyme A-C1; PLAAT1 |
| Gene Symbols | HRASLS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
