missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (Y13-238) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£209.00 - £486.00
Specifications
| Antigen | HRAS |
|---|---|
| Clone | Y13-238 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608454
|
Novus Biologicals
NBP3-20119-0.025mg |
0.025 mg |
£209.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673634
|
Novus Biologicals
NBP3-20119-0.2mg |
0.2 mg |
£486.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HRAS Monoclonal antibody specifically detects HRAS in Human, Mouse samples. It is validated for ImmunoprecipitationSpecifications
| HRAS | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Injection of rats subcutaneously with Ha-MuSV transformed NRK cells and screening of sera of individual animals to identify those with the strongest immune response against p21. Spleens were removed from the immunised animals and cell fusion of immune lymphocytes with rat myeloma cell line 210RCY3 Ag 1.2.3 (Y3) was carried out. Supernatants were harvested and assayed for antibodies to p21 by GDP binding assay or immunofluorescence on Ha-SV-transformed MDCK cells, followed by testing for the ability of the antibodies to immunoprecipitate p21. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| Y13-238 | |
| Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| PBS | |
| 3265 | |
| IgG2a λ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title