missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (H-Ras-03), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
£402.00
Specifications
| Antigen | HRAS |
|---|---|
| Clone | H-Ras-03 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 646 |
Description
HRAS Monoclonal antibody specifically detects HRAS in Human samples. It is validated for Western BlotSpecifications
| HRAS | |
| Western Blot | |
| Janelia Fluor 646 | |
| Mouse | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 50mM Sodium Borate | |
| 3265 | |
| IgG1 | |
| Protein A purified |
| H-Ras-03 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Peptide corresponding to amino acids DIHQYREQIKRVKDSDDC of human H-Ras protein. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title