missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Antibody (H-Ras-03), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68799R
This item is not returnable.
View return policy
Description
HRAS Monoclonal antibody specifically detects HRAS in Human samples. It is validated for Western Blot
Specifications
| HRAS | |
| Monoclonal | |
| DyLight 550 | |
| P01112 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| H-Ras-03 | |
| 50mM Sodium Borate | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Peptide corresponding to amino acids DIHQYREQIKRVKDSDDC of human H-Ras protein. | |
| 0.1 mL | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction