missing translation for 'onlineSavingsMsg'
Learn More

HR6B/UBE2B Antibody (PCRP-UBE2B-1C7) - Azide and BSA Free, Novus Biologicals™

Product Code. 18351453 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351453 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351453 Supplier Novus Biologicals Supplier No. NBP308912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HR6B/UBE2B Monoclonal specifically detects HR6B/UBE2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HR6B/UBE2B
Applications Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-UBE2B-1C7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B
Host Species Mouse
Immunogen Recombinant full-length human HR6B/UBE2B protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7320
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.