missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR6B/UBE2B Antibody (OTI1E11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71337UV
This item is not returnable.
View return policy
Description
HR6B/UBE2B Monoclonal specifically detects HR6B/UBE2B in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HR6B/UBE2B | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBE2B | |
| Full length recombinant protein of human HR6B/UBE2B (NP_003328) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| OTI1E11 | |
| Western Blot | |
| E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins | |
| 7320.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction