missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR6B/UBE2B Antibody (OTI1E11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71337CL1
This item is not returnable.
View return policy
Description
HR6B/UBE2B Monoclonal antibody specifically detects HR6B/UBE2B in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| HR6B/UBE2B | |
| Monoclonal | |
| CoraFluor 1 | |
| E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
| Full length human recombinant protein of human UBE2B (NP_003328) produced in E.coli. | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 7320 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot | |
| OTI1E11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction