missing translation for 'onlineSavingsMsg'
Learn More

HR6A/UBE2A Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-554AF532

Product Code. 18723401

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HR6A/UBE2A Polyclonal antibody specifically detects HR6A/UBE2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HR6A/UBE2A
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 6.3.2.19, hHR6A, HR6A, RAD6ARAD6 homolog A, UBC2, Ubiquitin carrier protein A, ubiquitin-conjugating enzyme E2 A, ubiquitin-conjugating enzyme E2A (RAD6 homolog), Ubiquitin-protein ligase A
A synthetic peptide which maps to a region between residues 10 and 50 of human ubiquitin-conjugating enzyme E2A using the numbering given in Swiss-Prot entry P49459 (GeneID 7319).
0.1 mL
DNA Repair, Ubiquitin Proteasome Pathway
7319
Store at 4C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.