missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR23A/Rad23A TUBE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | HR23A/Rad23A TUBE1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652198
|
Novus Biologicals
NBP2-68598-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658966
|
Novus Biologicals
NBP2-68598 |
100 μg |
£435.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HR23A/Rad23A TUBE1 Polyclonal antibody specifically detects HR23A/Rad23A TUBE1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| HR23A/Rad23A TUBE1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| hHR23A, HR23A | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LSDDVPIRDYRIDEKNFVVVMVTKTKAGQGTSAPPEASPTAAPESSTSFPPAPTSGMSHPPPAAREDKSPSEESAPTTSPESVSGSVPSSGSSGREEDAASTLVTGSEYETMLTEIMSMGYER | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 5886 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title