missing translation for 'onlineSavingsMsg'
Learn More

HQ-415, MedChemExpress

Product Code. 16447506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447506 100 mg 100mg
16483587 50 mg 50mg
16477568 10 mg 10mg
16440048 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447506 Supplier Medchem Express Supplier No. HY18670
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HQ-415 is a class of clinically relevant bioactive metal chelators related to clioquinol.

HQ-415 is a class of clinically relevant bioactive metal chelators related to clioquinol.

Specifications

Chemical Name or Material HQ-415
CAS 430462-93-4
Color White
Purity Grade Notes Research
Molecular Formula C25H25N3O3
Quantity 100 mg
Solubility Information DMSO : 5.6 mg/mL (13.48 mM; Need ultrasonic and warming)
SMILES OC1=C2N=CC=CC2=CC=C1C(C3=CC=C(OC)C(OCC)=C3)NC4=NC=CC(C)=C4
Molecular Weight (g/mol) 415.48
Formula Weight 415.48
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.