missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13223
This item is not returnable.
View return policy
Description
HPV18 E7 Polyclonal antibody specifically detects HPV18 E7 in Virus samples. It is validated for Western Blot, Immunofluorescence
Specifications
| HPV18 E7 | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| E7 protein | |
| Full length HPV18 E7 recombinant protein. | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction