missing translation for 'onlineSavingsMsg'
Learn More

HPV18 E7 Antibody (GT881), Novus Biologicals™

Product Code. 18333253
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333253 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333253 Supplier Bio-Techne Supplier No. NBP313610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HPV18 E7 Monoclonal antibody specifically detects HPV18 E7 in Virus samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV18 E7
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT881
Conjugate Unconjugated
Formulation PBS
Gene Alias E7 protein
Host Species Mouse
Immunogen Full length HPV18 E7 recombinant protein.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.