missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (HPV18/1297), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08845AF647
This item is not returnable.
View return policy
Description
HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HPV18 E6 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| HPV18 E6-beta-galactosidase fusion protein | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| Virus | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HPV18/1297 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction