missing translation for 'onlineSavingsMsg'
Learn More

HPV18 E6 Antibody (C1N1), DyLight 594, Novus Biologicals™

Product Code. 18402053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402053 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402053 Supplier Novus Biologicals Supplier No. NBP254807DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV18 E6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone C1N1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
Host Species Mouse
Immunogen HPV18E6-beta-galactosidase fusion protein synthesized in E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.