missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E2 Mouse anti-Virus, Clone: TVG 261, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | HPV16 E2 |
|---|---|
| Clone | TVG 261 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL |
| Applications | Western Blot |
Description
HPV16 E2 Monoclonal specifically detects HPV16 E2 in Virus samples. It is validated for Western Blot.Specifications
| HPV16 E2 | |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| HPV16 regulatory protein E2, Human Papilloma Virus 16 E2, Human papilloma virus 16 regulatory protein E2, Human papillomavirus type 16 E2, Human papillomavirus type 16 regulatory protein E2, Regulatory protein E2 | |
| IgG1 κ | |
| Protein A or G purified |
| TVG 261 | |
| Western Blot 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Raised against Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title