missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPS6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530827
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (85%), Rat (85%), Pig (86%), Rhesus Monkey (96%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6.
Specifications
| HPS6 | |
| Polyclonal | |
| Unconjugated | |
| Hps6 | |
| 5330434M19Rik; BLOC2; BLOC2S3; FLJ22501; Hermansky-Pudlak syndrome 6; Hermansky-Pudlak syndrome 6 protein; Hermansky-Pudlak syndrome 6 protein homolog; Hermansky-Pudlak syndrome-6 protein (HPS6); Hps6; HPS6, biogenesis of lysosomal organelles complex 2 subunit 3; Hsp6; MGC20522; RP11-302K17.1; ru; ruby eye; ruby-eye protein; Ruby-eye protein homolog; ruby-eye-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.95 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q86YV9 | |
| Hps6 | |
| Recombinant protein encompassing a sequence within the center region of human HPS6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction