missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33681
This item is not returnable.
View return policy
Description
HPS6 Polyclonal specifically detects HPS6 in Human samples. It is validated for Western Blot.
Specifications
| HPS6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| HPS6 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human HPS6. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79803 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| FLJ22501, Hermansky-Pudlak syndrome 6, Hermansky-Pudlak syndrome 6 protein, Hermansky-Pudlak syndrome-6 protein (HPS6), MGC20522, RP11-302K17.1, Ru, Ruby-eye protein homolog | |
| Rabbit | |
| 83 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction