Learn More
Invitrogen™ HPS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588334
Description
Immunogen sequence: IISSRRGSQS DEDSCSLHSQ TLSEDERFKE FTSQQEEDLP DQCCGSHGNE DNVSHAPVMF ETDKNETFLP FGIPLPFRSP SPLVSLQAVK ESVSSFVRKT TEKIGTLHTS PDLKVRPELR GDEQSCEEDV SSDTCPKEED TEEEKEVTSP PPEEDRFQEL KVATAEAMTK LQDPLVLFES ESLRMVLQEW LSHLEKTFAM KDFSGVSDTD NSSMKLNQDV LLVNESKKGI LDEDNEKEKR DSLGNEESVD KTACECVRSP R; Positive Samples: Jurkat, 293T, B cells; Cellular Location: Cytoplasm, cytosol.
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.
Specifications
| HPS5 | |
| Polyclonal | |
| Unconjugated | |
| Hps5 | |
| Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; alpha integrin binding protein 63; alpha-integrin-binding protein 63; BLOC2S2; C85120; haze; Hermansky-Pudlak syndrome 5; Hermansky-Pudlak syndrome 5 homolog (human); Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; HPS5; HPS5, biogenesis of lysosomal organelles complex 2 subunit 2; hz; KIAA1017; maroon; mr; Ru2; ru-2; ruby eye 2; ruby-eye protein 2; Ruby-eye protein 2 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UPZ3 | |
| Hps5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-600 of human HPS5 (NP_0091473). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.