missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100099
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HPS5.
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.
Specifications
| HPS5 | |
| Polyclonal | |
| Unconjugated | |
| Hps5 | |
| Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; alpha integrin binding protein 63; alpha-integrin-binding protein 63; BLOC2S2; C85120; haze; Hermansky-Pudlak syndrome 5; Hermansky-Pudlak syndrome 5 homolog (human); Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; HPS5; HPS5, biogenesis of lysosomal organelles complex 2 subunit 2; hz; KIAA1017; maroon; mr; Ru2; ru-2; ruby eye 2; ruby-eye protein 2; Ruby-eye protein 2 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11234, 246694, 308598 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59438, Q9UPZ3 | |
| Hps5 | |
| A synthesized peptide derived from human HPS5(Accession Q9UPZ3), corresponding to amino acid residues R435-T485. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction