missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14627-1-AP-20UL
This item is not returnable.
View return policy
Description
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintece of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).Specifications
| HPS4 | |
| Polyclonal | |
| Unconjugated | |
| HPS4 | |
| bK1048E9.4, bK1048E9.5, Hermansky Pudlak syndrome 4, HPS4, KIAA1667, LE, Light ear protein homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 89781 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NQG7 | |
| HPS4 | |
| HPS4 Fusion Protein Ag6202 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction