missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562152
This item is not returnable.
View return policy
Description
Immunogen sequence: GQRGNKLPTG EQGLDEDVDG VCESHAAPGL ECSSGSANCQ GAGPSADGIS SRLTPAESCM GLVRMNLYTH CVKGLVLSLL AEEPLLGDSA AIEEVYHSSL ASLNGLEVHL KETLPRDEAA STSSTYNFTH YDR Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 58%.
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Specifications
| HPS4 | |
| Polyclonal | |
| Unconjugated | |
| HPS4 | |
| 2010205O06Rik; AU040608; BLOC-3; BLOC3S2; C130020P05Rik; Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 homolog; Hermansky-Pudlak syndrome 4 homolog (human); Hermansky-Pudlak syndrome 4 protein; Hermansky-Pudlak syndrome 4 protein homolog; Hps4; HPS4, biogenesis of lysosomal organelles complex 3 subunit 2; KIAA1667; Le; Le protein; light ear; light-ear protein; Light-ear protein homolog; mKIAA1667 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89781 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQG7 | |
| HPS4 | |
| Recombinant protein corresponding to Human HPS4. Recombinant protein control fragment (Product #RP-91492). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction