missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPRT Rabbit anti-Human, Mouse, Rat, Clone: 4N2Q6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | HPRT |
|---|---|
| Clone | 4N2Q6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327343
|
Novus Biologicals
NBP3-16702-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18368311
|
Bio-Techne
NBP3-16702-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPRT Monoclonal antibody specifically detects HPRT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HPRT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3251 | |
| IgG | |
| Affinity purified |
| 4N2Q6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
| A synthetic peptide corresponding to a sequence within amino acids 119-218 of human HPRT1 (P00492). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel