missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HPRT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29542
This item is not returnable.
View return policy
Description
HPRT Polyclonal antibody specifically detects HPRT in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| HPRT | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
| Between 168 and 218 | |
| 100 μg | |
| Lipid and Metabolism | |
| 3251 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction