missing translation for 'onlineSavingsMsg'
Learn More

HPOB, MedChemExpress

Product Code. 16440348
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440348

Brand: Medchem Express HY19747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HPOB is a highly potent and selective inhibitor of HDAC6 with an IC50 of 56 nM. HPOB displays >30 fold less potent against other HDACs. HPOB enhances the effectiveness of DNA-damaging anticancer agents in transformed cells but not normal cells. HPOB does not block the ubiquitin-binding activity of HDAC6.

HPOB is a highly potent and selective inhibitor of HDAC6 with an IC50 of 56 nM. HPOB displays >30 fold less potent against other HDACs. HPOB enhances the effectiveness of DNA-damaging anticancer agents in transformed cells but not normal cells. HPOB does not block the ubiquitin-binding activity of HDAC6.

Specifications

Chemical Name or Material HPOB
Physical Form Solid
CAS 1429651-50-2
Color Pink
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H18N2O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (159.06 mM; Need ultrasonic)
SMILES O=C(N(CCO)C1=CC=CC=C1)CC2=CC=C(C(NO)=O)C=C2
Molecular Weight (g/mol) 314.34
Formula Weight 314.34
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.