Learn More
SCAT HPLC Supply-Sets, V2.0
The HPLC Supply Sets, V2.0 of the manufacturer SCAT are combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as capillaries, suction filters and air valves.
Brand:  SCAT 307347
144.92 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.78000kg
 View more versions of this product
Product Code. 16453799
  • £171.00 / 1 set
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Specifications
Specifications
1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle Youtility & clear glass + 3 x 1.5 m capillaries 3.2 mm OD, 3 x PTFE suction filters + air valve (317010)
Clear
GL45
Set III
Any HPLC System
HPLC Supply-Set III, V2.0, 1 x Safety Cap III GL45, 1x 1000 mL Laboratory bottle, Youtility & clear glass + 3 x 1,5 m capillary 3,2 mm OD, 3 x PTFE suction filter + air valve (317010)