missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Be prepared for HPLC with these SCAT™ suction filters.
Specifications
Specifications
| For Use With (Equipment) | HPLC Instrument |
| Material | UHMW-PE |
| Pore Size | 20 μm |
| Type | HPLC Suction Filter |
| Diameter (Metric) Outer | 3.2 mm |
| Quantity | 5 Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?