missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ HPLC Safety Cap II
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
£110.56
Specifications
Autoclavable | Autoclavable |
---|---|
Type | Safety Cap |
For Use With (Application) | HPLC Systems |
For Use With (Equipment) | Lab Bottles |
Includes | 2 PFA Fittings with 1.6/2.3/3.2 mm OD, 1 PFA Blind Plugs, 1 Air Valve |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
17967260
|
SCAT™
107511 |
GL32 |
£110.56
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17907250
|
SCAT™
107637 |
38-430 |
£110.56
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every SafetyCap
Specifications
Autoclavable | |
HPLC Systems | |
2 PFA Fittings with 1.6/2.3/3.2 mm OD, 1 PFA Blind Plugs, 1 Air Valve |
Safety Cap | |
Lab Bottles | |
PTFE |