missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HPLC Qualification Kits for UltiMate™ 3000 Systems

Product Code. 17881256 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Qualification Kit (OQ/PQ) - Manual Injector
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17881256 Qualification Kit (OQ/PQ) - Manual Injector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17881256 Supplier Thermo Scientific™ Supplier No. 4832.5000A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HPLC Qualification Kits are for system validation of UltiMate 3000 LC and UHPLC systems.

Choose Thermo Scientific™ Dionex™ UltiMate™ HPLC Qualification Kits for system validation of UltiMate 3000 LC and UHPLC systems.

The modules of UltiMate 3000 RSLCnano system can be arranged in different configurations for various applications.
  • Components include fittings, capillaries and columns, but vary by application

Specifications

Description Qualification Kit (OQ/PQ) - Manual Injector
For Use With (Equipment) Dionex UltiMate 3000 Series
Product Line UltiMate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.