Learn More
Macherey Nagel HPLC guard column EC 4/2NUCLEODUR 100-5 NH2-RP lenght: 4 mm,
Brand:  Macherey Nagel 761953.20
HPLC guard column EC 4/2NUCLEODUR 100-5 NH2-RP lenght: 4 mm,
Product Code. 12788982
  • £141.00 / Pack of 1
Only null left
Estimated Shipment: 27-05-2026