Learn More
VICI Jour Chromatography™ HPLC Fitting Kit
Specifications
Specifications
| For Use With (Application) | NO-OX tubing |
| Includes | PCTFE tubing inserts, 1/8 in. PPS nuts, 1/8 in. ETFE ferrules |
| Material | PCTFE |